Merck
  • Total synthesis and molecular target of largazole, a histone deacetylase inhibitor.
Total synthesis and molecular target of largazole, a histone deacetylase inhibitor.
Journal of the American Chemical Society (2008-05-30)
Yongcheng Ying, Kanchan Taori, Hyoungsu Kim, Jiyong Hong, Hendrik Luesch
ABSTRACT
Full details of the concise and convergent synthesis (eight steps, 19% overall yield), its extension to the preparation of a series of key analogues, and the molecular target and pharmacophore of largazole are described. Central to the synthesis of largazole is a macrocyclization reaction for formation of the strained 16-membered depsipeptide core followed by an olefin cross-metathesis reaction for installation of the thioester. The biological evaluation of largazole and its key analogues, including an acetyl analogue, a thiol analogue, and a hydroxyl analogue, suggested that histone deacetylases (HDACs) are molecular targets of largazole and largazole is a class I HDAC inhibitor. In addition, structure-activity relationship (SAR) studies revealed that the thiol group is the pharmacophore of the natural product. Largazole's HDAC inhibitory activity correlates with its antiproliferative activity.
Sigma-Aldrich
4-Bromo-1-butene, 97%