Merck
  • Synthesis of 2,2'-bipyrroles and 2,2'-thienylpyrroles from donor-acceptor cyclopropanes and 2-cyanoheteroles.
Synthesis of 2,2'-bipyrroles and 2,2'-thienylpyrroles from donor-acceptor cyclopropanes and 2-cyanoheteroles.
Organic letters (2004-03-12)
Ming Yu, G Dan Pantos, Jonathan L Sessler, Brian L Pagenkopf
ABSTRACT
[reaction: see text] Two new series of 2,2'-bipyrroles and 2,2'-thienylpyrroles have been prepared by trimethylsilyl trifluoromethanesulfonate (TMSOTf)-mediated reaction of donor-acceptor cyclopropanes with 2-cyanopyrroles and 2-cyanothiophene, respectively. This method opens the door toward a wide variety of unsymmetrical bipyrroles and thienylpyrroles.
Sigma-Aldrich
2-Thiophenecarbonitrile, 99%