Merck
  • Synthesis of 2,6-dioxo-(1H,3H)-9-N-ribitylpurine and 2,6-dioxo-(1H,3H)-8-aza-9-N-ribitylpurine as inhibitors of lumazine synthase and riboflavin synthase.
Synthesis of 2,6-dioxo-(1H,3H)-9-N-ribitylpurine and 2,6-dioxo-(1H,3H)-8-aza-9-N-ribitylpurine as inhibitors of lumazine synthase and riboflavin synthase.
Bioorganic & medicinal chemistry (1998-05-23)
M Cushman, F Mavandadi, K Kugelbrey, A Bacher
ABSTRACT
2,6-Dioxo-(1H,3H)-9-N-ribitylpurine (6) and 2,6-dioxo-(1H,3H)-8-aza-9-N-ribitylpurine (7) have been synthesized and evaluated as inhibitors of lumazine synthase and riboflavin synthase. Reaction of 5-amino-6-ribitylaminouracil hydrochloride (8) with diethoxymethyl acetate (9) afforded the purine 6, while diazotization of 8 afforded the 8-aza purine 7. Compounds 6 and 7 were evaluated against lumazine synthase of Bacillus subtilis and riboflavin synthase of Escherichia coli. Both 6 and 7 were better inhibitors of lumazine synthase than riboflavin synthase. The 8-azapurine 7 had a lower KI (0.33 and 0.39 mM) than the purine 6 (0.47 and 0.54 mM) when evaluated with lumazine synthase and riboflavin synthase, respectively.
Sigma-Aldrich
Diethoxymethyl acetate, 96%