Merck
  • CAMP (C13orf8, ZNF828) is a novel regulator of kinetochore-microtubule attachment.
CAMP (C13orf8, ZNF828) is a novel regulator of kinetochore-microtubule attachment.
The EMBO journal (2010-11-11)
Go Itoh, Shin-ichiro Kanno, Kazuhiko S K Uchida, Shuhei Chiba, Shiro Sugino, Kana Watanabe, Kensaku Mizuno, Akira Yasui, Toru Hirota, Kozo Tanaka
ABSTRACT
Proper attachment of microtubules to kinetochores is essential for accurate chromosome segregation. Here, we report a novel protein involved in kinetochore-microtubule attachment, chromosome alignment-maintaining phosphoprotein (CAMP) (C13orf8, ZNF828). CAMP is a zinc-finger protein containing three characteristic repeat motifs termed the WK, SPE, and FPE motifs. CAMP localizes to chromosomes and the spindle including kinetochores, and undergoes CDK1-dependent phosphorylation at multiple sites during mitosis. CAMP-depleted cells showed severe chromosome misalignment, which was associated with the poor resistance of K-fibres to the tension exerted upon establishment of sister kinetochore bi-orientation. We found that the FPE region, which is responsible for spindle and kinetochore localization, is essential for proper chromosome alignment. The C-terminal region containing the zinc-finger domains negatively regulates chromosome alignment, and phosphorylation in the FPE region counteracts this regulation. Kinetochore localization of CENP-E and CENP-F was affected by CAMP depletion, and by expressing CAMP mutants that cannot functionally rescue CAMP depletion, placing CENP-E and CENP-F as downstream effectors of CAMP. These data suggest that CAMP is required for maintaining kinetochore-microtubule attachment during bi-orientation.