Merck
  • Microbial transformation of pharmaceuticals naproxen, bisoprolol, and diclofenac in aerobic and anaerobic environments.
Microbial transformation of pharmaceuticals naproxen, bisoprolol, and diclofenac in aerobic and anaerobic environments.
Archives of environmental contamination and toxicology (2010-11-18)
Marja Lahti, Aimo Oikari
ABSTRACT
Although biotransformation is generally considered to be the main process by which to remove pharmaceuticals, both in sewage treatment plants and in aquatic environments, quantitative information on specific compounds is scarce. In this study, the transformations of diclofenac (DCF), naproxen (NPX), and bisoprolol (BSP) were studied under aerobic and anaerobic conditions using inocula taken from activated and digested sludge processes, respectively. Whereas concentration decays were monitored by LC-tandem mass spectrometry, oxygen consumption and methane production were used for the evaluation of the performance of overall conditions. DCF was recalcitrant against both aerobic and anaerobic biotransformation. More than one third of the BSP disappeared under aerobic conditions, whereas only 14% was anaerobically biotransformed in 161 days. Under aerobic conditions, complete removal of NPX was evident within 14 days, but anaerobic transformation was also efficient. Formation of 6-O-desmethylnaproxen, a previously reported aerobic metabolite, was also detected under anaerobic conditions and persisted for 161 days.
Sigma-Aldrich
N-Allylthiourea, 98%