Merck
  • A new method for the determination of L-dopa and 3-O-methyldopa in plasma and cerebrospinal fluid using gas chromatography and electron capture negative ion mass spectrometry.
A new method for the determination of L-dopa and 3-O-methyldopa in plasma and cerebrospinal fluid using gas chromatography and electron capture negative ion mass spectrometry.
Clinica chimica acta; international journal of clinical chemistry (1988-01-15)
A P de Jong, R M Kok, C A Cramers, S K Wadman, E Haan
ABSTRACT
L-3-(3,4-Dihydroxyphenyl)alanine (DOPA) and its 3-O-methyl metabolite (OMD) were measured in plasma and cerebrospinal fluid by a new assay which combines N,O-acetylation of amino acids in aqueous media, preparation of pentafluorobenzyl esters under anhydrous conditions, and analysis by gas chromatography-electron capture negative ion mass spectrometry. The N,O-acetyl, carboxy-PFB derivatives gave abundant carboxylate anions ([M-CH2C6F5]-) which were suitable for sensitive analysis using selected ion monitoring. Plasma and CSF samples were sufficiently purified by a simple organic solvent extraction. Analytical recovery for DOPA was 100.2 +/- 3.7% at the level of 100 nmol/l. Analysis of DOPA in plasma was performed with a relative standard deviation of 5%. The limit of quantitation in plasma and CSF was at the sub-nmol/l level. In healthy adults, DOPA concentration in plasma was 9.0 +/- 2 nmol/l (n = 11) and in CSF 3.5 +/- 0.9 nmol/l (n = 9). The concentration of OMD in plasma was 99.1 nmol/l (pool of 24 samples) and 15.3 nmol/l in CSF (pool of 12 samples). Measurement of 5-[2H]DOPA and 5-[2H]OMD in plasma of a healthy individual who had been orally loaded with 3,5-[2H2]tyrosine (150 mg kg body wt) was possible for several hours after the load.
Product Description
Supelco
Levodopa, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, Vetec, reagent grade, 98%