Merck
  • Noxious cold ion channel TRPA1 is activated by pungent compounds and bradykinin.
Noxious cold ion channel TRPA1 is activated by pungent compounds and bradykinin.
Neuron (2004-03-30)
Michael Bandell, Gina M Story, Sun Wook Hwang, Veena Viswanath, Samer R Eid, Matt J Petrus, Taryn J Earley, Ardem Patapoutian
ABSTRACT
Six members of the mammalian transient receptor potential (TRP) ion channels respond to varied temperature thresholds. The natural compounds capsaicin and menthol activate noxious heat-sensitive TRPV1 and cold-sensitive TRPM8, respectively. The burning and cooling perception of capsaicin and menthol demonstrate that these ion channels mediate thermosensation. We show that, in addition to noxious cold, pungent natural compounds present in cinnamon oil, wintergreen oil, clove oil, mustard oil, and ginger all activate TRPA1 (ANKTM1). Bradykinin, an inflammatory peptide acting through its G protein-coupled receptor, also activates TRPA1. We further show that phospholipase C is an important signaling component for TRPA1 activation. Cinnamaldehyde, the most specific TRPA1 activator, excites a subset of sensory neurons highly enriched in cold-sensitive neurons and elicits nociceptive behavior in mice. Collectively, these data demonstrate that TRPA1 activation elicits a painful sensation and provide a potential molecular model for why noxious cold can paradoxically be perceived as burning pain.
Sigma-Aldrich
Cinnamaldehyde, Vetec, reagent grade, 93%