Merck
  • Relative binding of certain steroids of low polarity to human sex hormone-binding globulin: strong binding of 2-methoxyestrone, a steroid lacking the 17 beta-OH group.
Relative binding of certain steroids of low polarity to human sex hormone-binding globulin: strong binding of 2-methoxyestrone, a steroid lacking the 17 beta-OH group.
Steroids (1986-06-01)
A Philip, B E Murphy
ABSTRACT
The cross-reactivities for binding sites on human sex hormone-binding globulin (SHBG) of a number of steroids of low polarity, including progesterone and estrogen metabolites, were studied. Binding relative to testosterone (100%) was low (less than or equal to 3%) except for 2-methoxyestrone (81%), 3-acetoxyestradiol (16%), 4-methoxyestradiol (6%), 3 beta-hydroxy-5 beta-pregnan-3-one (5.8%), 5 alpha-dihydrodeoxycorticosterone (4.5%), deoxycorticosterone (4%), 20 alpha-hydroxy-5 alpha-pregnan -3-one (4%), 4-methoxyestrone (4%) and 5 alpha-dihydroprogesterone (3.5%). 2-Methoxyestrone is the only steroid lacking a 17 beta-hydroxyl group which binds to SHBG with strong affinity.