Merck
  • The impairment of HCCS leads to MLS syndrome by activating a non-canonical cell death pathway in the brain and eyes.
The impairment of HCCS leads to MLS syndrome by activating a non-canonical cell death pathway in the brain and eyes.
EMBO molecular medicine (2012-12-15)
Alessia Indrieri, Ivan Conte, Giancarlo Chesi, Alessia Romano, Jade Quartararo, Rosarita Tatè, Daniele Ghezzi, Massimo Zeviani, Paola Goffrini, Ileana Ferrero, Paola Bovolenta, Brunella Franco
ABSTRACT
Mitochondrial-dependent (intrinsic) programmed cell death (PCD) is an essential homoeostatic mechanism that selects bioenergetically proficient cells suitable for tissue/organ development. However, the link between mitochondrial dysfunction, intrinsic apoptosis and developmental anomalies has not been demonstrated to date. Now we provide the evidence that non-canonical mitochondrial dependent apoptosis explains the phenotype of microphthalmia with linear skin lesions (MLS), an X-linked developmental disorder caused by mutations in the holocytochrome c-type synthase (HCCS)gene [corrected]. By taking advantage of a medaka model that recapitulates the MLS phenotype we demonstrate that downregulation of hccs, an essential player of the mitochondrial respiratory chain (MRC), causes increased cell death via an apoptosome-independent caspase-9 activation in brain and eyes. We also show that the unconventional activation of caspase-9 occurs in the mitochondria and is triggered by MRC impairment and overproduction of reactive oxygen species (ROS). We thus propose that HCCS plays a key role in central nervous system (CNS) development by modulating a novel non-canonical start-up of cell death and provide the first experimental evidence for a mechanistic link between mitochondrial dysfunction, intrinsic apoptosis and developmental disorders.
Sigma-Aldrich
[D-Trp7, Ala8, D-Phe10]-α-Melanocyte Stimulating Hormone Amide Fragment 6-11, ≥97% (HPLC)