- Therapeutic drug monitoring of thiopurine metabolites in adult thiopurine tolerant IBD patients on maintenance therapy.
Therapeutic drug monitoring of thiopurine metabolites in adult thiopurine tolerant IBD patients on maintenance therapy.
Therapeutic drug monitoring of active metabolites of thiopurines, azathioprine and 6-mercaptopurine, is relatively new. The proposed therapeutic threshold level of the active 6-thioguanine nucleotides (6-TGN) is ≥235 pmol/8×10(8) erythrocytes. The aim of this prospective cross-sectional study was to compare 6-TGN levels in adult thiopurine tolerant IBD patients with an exacerbation with those in remission, and to determine the therapeutic 6-TGN cut-off level. Hundred IBD patients were included. Outcome measures were thiopurine metabolite levels, calculated therapeutic 6-TGN cut-off level, CDAI/CAI scores, thiopurine dose and TPMT enzyme activity. Forty-one patients had an exacerbation, 59 patients were in remission. In 17% of all patients 6-TGN levels were compatible with non-compliance. The median 6-TGN levels were not significantly different between the exacerbation and remission group (227 versus 263 pmol/8×10(8) erythrocytes, p=0.29). The previous reported therapeutic 6-TGN cut-off level of 235 pmol/8×10(8) erythrocytes was confirmed in this study. Twenty-six of the 41 patients (63%) with active disease had 6-TGN levels below this threshold and 24 of 59 IBD patients (41%) in clinical remission (p=0.04). Thiopurine non-compliance occurs frequently both in active and quiescent disease. 6-TGN levels below or above the therapeutic threshold are associated with a significant higher chance of IBD exacerbation and remission, respectively. These data support the role of therapeutic drug monitoring in thiopurine maintenance therapy in IBD to reveal non-compliance or underdosing, and can be used as a practical tool to optimize thiopurine therapy, especially in case of thiopurine non-response.