Merck
  • Mechanism of photooxidation of folic acid sensitized by unconjugated pterins.
Mechanism of photooxidation of folic acid sensitized by unconjugated pterins.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2010-10-06)
M Laura Dántola, M Paula Denofrio, Beatriz Zurbano, Carlos S Gimenez, Peter R Ogilby, Carolina Lorente, Andrés H Thomas
ABSTRACT
Folic acid, or pteroyl-l-glutamic acid (PteGlu), is a precursor of coenzymes involved in the metabolism of nucleotides and amino acids. PteGlu is composed of three moieties: a 6-methylpterin (Mep) residue, a p-aminobenzoic acid (PABA) residue, and a glutamic acid (Glu) residue. Accumulated evidence indicates that photolysis of PteGlu leads to increased risk of several pathologies. Thus, a study of PteGlu photodegradation can have significant ramifications. When an air-equilibrated aqueous solution of PteGlu is exposed to UV-A radiation, the rate of the degradation increases with irradiation time. The mechanism involved in this "auto-photo-catalytic" effect was investigated in aqueous solutions using a variety of tools. Whereas PteGlu is photostable under anaerobic conditions, it is converted into 6-formylpterin (Fop) and p-aminobenzoyl-l-glutamic acid (PABA-Glu) in the presence of oxygen. As the reaction proceeds and enough Fop accumulates in the solution, a photosensitized electron-transfer process starts, where Fop photoinduces the oxidation of PteGlu to Fop, and H(2)O(2) is formed. This process also takes place with other pterins as photosensitizers. The results are discussed with the context of previous mechanisms for processes photosensitized by pterins, and their biological implications are evaluated.
Sigma-Aldrich
N-(4-Aminobenzoyl)-L-glutamic acid, ≥98% (TLC)
Folic acid impurity A, European Pharmacopoeia (EP) Reference Standard