Merck
  • Chronic actinic dermatitis to sesquiterpene lactones: [2+2] photoreaction toward thymidine of (+) and (-) alpha-methylene-hexahydrobenzofuranone with a cis ring junction.
Chronic actinic dermatitis to sesquiterpene lactones: [2+2] photoreaction toward thymidine of (+) and (-) alpha-methylene-hexahydrobenzofuranone with a cis ring junction.
Photochemistry and photobiology (2010-02-02)
Sébastien Fuchs, Valérie Berl, Jean-Pierre Lepoittevin
ABSTRACT
(+) and (-) alpha-methylene-hexahydrobenzofuranone derivatives with a stereochemically pure cis ring junction were used as models of sesquiterpene lactones to study their photoreactivity toward thymidine. After 313 nm irradiation of a deoxygenated acetone solution of lactone models and thymidine, six [2+2] photoadducts were isolated for each enantiomer and fully characterized by a combination of NMR experiments. A common syn regioselectivity and exo stereoselectivity were observed for photoadducts. This high photoreactivity of alpha-methylene-gamma-butyrolactone ring toward thymidine could be an explanation of the progressive evolution of allergic contact dermatitis toward chronic actinic dermatitis.
Sigma-Aldrich
α-Methylene-γ-butyrolactone, 97%