Merck
[Study of cytotoxic and antiviral effects of some eye drops].
Oftalmologia (Bucharest, Romania : 1990) (2008-01-01)
Lucreţia Dediulescu, Daniela Florentina Dediulescu
ABSTRACT
The study of the cytotoxic and antiviral effect of six commercial mixtures, eye drops type, underlined the advantages of using eye drops with Indomethacin for Herpetic Keratitis, due to the antiviral effect and also for the lack of cytotoxicity.