Merck
  • [Treatment in cases of recurrent vaginal candida infection during pregnancy].
[Treatment in cases of recurrent vaginal candida infection during pregnancy].
Akusherstvo i ginekologiia (2008-07-23)
A Nikolov, E Shopova, M Mikhova, A Dimitrov
ABSTRACT
To study the therapeutic application of Gynazol in cases of recurrent vaginal Candida infections during the last trimester of pregnancy. 28 pregnant women in third trimester with recurrent vaginal Candida infection were included in the study. All cases were treated with a single vaginal application of Gynazol (butoconazole nitrate 2%). Follow up of reported subjective complaints and control vaginal swab and culture 10-14 days after treatment was performed. In 24 (85.72%) cases of recurrent vaginal Candida infection, Candida albicans was isolated. C. tropicalis and C. parapsilosis were found in 3 (10.71%) and 1 (3.57%) cases respectively. Single vaginal application of Gynazol led to alleviation of subjective symptoms 24-48 hours after initiation of treatment. Control vaginal swab and culture were performed in 24 patients. Negative cultures were found in 21 cases (87.5%) and single Candida colonies were isolated in 3 (12.5%) patients. According to our results, application of Gynazol is recommended in cases of recurrent vaginal Candida infection during third trimester of pregnancy.