Merck
[Realization of optic activatory mechanism in laser-medication impact].
Voprosy kurortologii, fizioterapii, i lechebnoi fizicheskoi kultury (2006-06-07)
M Iu Gerasimenko, S N Skovorod'ko, O A Kondakova, A S Varavva, V F Barybin, V Ia Mendeleev
ABSTRACT
Clinical, biochemical, experimental, heat physical studies and mathematical calculations have shown that combined effect of drug-laser can be differentiated into the following effects: photophoresis, light pressure and photodynamic effect.
Sigma-Aldrich
6-Methyluracil, 97%