Merck
  • The antifouling activity of some juvenoids on three species of acorn barnacle, Balanus.
The antifouling activity of some juvenoids on three species of acorn barnacle, Balanus.
Pest management science (2006-05-18)
Lars Skattebøl, Nils Olav Nilsen, Yngve Stenstrøm, Petter Andreassen, Peter Willemsen
ABSTRACT
Analogues of insect juvenile hormones (juvenoids) have been tested for settling inhibition of cyprids from three species of barnacle, Balanus balanoides (L.), Balanus improvisus Darwin and Balanus amphitrite Darwin. Some 3-alkoxypyridine derivatives exhibited strong activity at mg litre(-1) concentrations; 3,7-dimethyloctyl 2-methyl-5-pyridyl ether (8) gave an EC(50) of 0.006 mg litre(-1) when tested on B. balanoides. When compound 8 and similar juvenoids were incorporated into paints, test panels kept in seawater for 8 months were free of barnacles.