- 3 Beta-(3,4-dihydroxycinnamoyl)-erythrodiol, a cytotoxic constituent of Zygophyllum geslini collected in the Algerian Sahara.
3 Beta-(3,4-dihydroxycinnamoyl)-erythrodiol, a cytotoxic constituent of Zygophyllum geslini collected in the Algerian Sahara.
Journal of ethnopharmacology (2004-10-28)
Dalila Smati, Arlette Longeon, Michèle Guyot
PMID15507367
ABSTRACT
3beta-(3,4-Dihydroxycinnamoyl)-erythrodiol was isolated as the cytotoxic constituent of the roots of Zygophyllum geslini.