Merck
  • Structural properties of cyclic peptides containing cis- or trans-2-aminocyclohexane carboxylic acid.
Structural properties of cyclic peptides containing cis- or trans-2-aminocyclohexane carboxylic acid.
Organic & biomolecular chemistry (2004-04-06)
Ulf Strijowski, Norbert Sewald
ABSTRACT
A series of cyclic peptides containing either cis- or trans-2-aminocyclohexane carboxylic acid as mimics for L-proline has been synthesized and their structural properties have been investigated using NMR and MD methods.