Merck
  • Convenient synthesis of 2'-deoxy-2-fluoroadenosine from 2-fluoroadenine.
Convenient synthesis of 2'-deoxy-2-fluoroadenosine from 2-fluoroadenine.
Nucleosides, nucleotides & nucleic acids (2003-11-12)
Song Ye, Martha M Rezende, Wei-Ping Deng, Kenneth L Kirk
ABSTRACT
A convenient synthesis of 2'-deoxy-2-fluoroadenosine from commercially available 2-fluoroadenine is described. The coupling reaction of silylated 2-fluoroadenine with phenyl 3,5-bis[O-(t-butyldimethylsilyl)]-2-deoxy-1-thio-D-erythro-pentofuranoside gave the corresponding 2-fluoro-2'-deoxyadenosine derivative (alpha/beta = 1:1) in good yield. The alpha- and beta-anomers were separated by chromatography, and then desilylated to give compounds 1a and 1b.
Sigma-Aldrich
2-Fluoroadenine, 96%