Merck
  • Utilization of 3-chloro-2-methylbenzoic acid by Pseudomonas cepacia MB2 through the meta fission pathway.
Utilization of 3-chloro-2-methylbenzoic acid by Pseudomonas cepacia MB2 through the meta fission pathway.
Applied and environmental microbiology (1992-08-01)
F K Higson, D D Focht
ABSTRACT
Pseudomonas cepacia MB2 grew on 3-chloro-2-methylbenzoate as a sole carbon source by metabolism through the meta fission pathway with the subsequent liberation of chloride. meta pyrocatechase activity in cell extracts was induced strongly by 3-chloro-2-methylbenzoate, but not by nongrowth analogs 4- or 5-chloro-2-methylbenzoate. Although rapid turnover of metabolites precluded direct identification, a mutant strain MB2-G5 lacking meta pyrocatechase activity produced 4-chloro-3-methylcatechol when incubated with 3-chloro-2-methylbenzoate. The catecholic product, confirmed by nuclear magnetic resonance and mass spectral analyses, produced a transient meta fission product (lambda max = 391 nm) from cell extracts of the wild-type MB2 strain. Further confirmation of meta pyrocatechase activity was noted by conversion of 4-chlorocatechol to 2-hydroxy-5-chloromuconic semialdehyde, which was not further metabolized. In contrast to 3-chlorocatechol, which was not metabolized and is known to generate suicidal products, 4-chlorocatechols do not generate acyl halides. Thus, further metabolism of the ring fission products is governed in strain MB2 by their suitability as substrates for the hydrolase.