Merck
[Characterization of lipopolysaccharides from Ralstonia solanacearum].
Mikrobiologiia (2003-04-18)
L D Varbanets, V N Vasil'ev, O S Brovarskaia
ABSTRACT
Lipopolysaccharides (LPSs) from four strains of Ralstonia solanacearum belonging to biovar I (ICMP 6524, 8115, 5712, and 8169) were isolated and investigated. The structural components of the LPS molecule, such as lipid A, the core oligosaccharide, and O-specific polysaccharide (O-PS), were obtained after mild acid hydrolysis of the LPS preparations. In lipid A from all the LPS samples studied, 3-hydroxyhexadecanoic, 2-hydroxyhexadecanoic, tetradecanoic, and hexadecanoic fatty acids prevailed. The dominant monosaccharides of the core oligosaccharides of all of the strains studied were rhamnose, glucose, glucosamine, 2-keto-3-deoxyoctulosonic acid, and heptose. However, individual strains varied in the content of galactose, ribose, xylose, and arabinose. Three types of the O-PS structure were established, which differed in their configuration (alpha or beta), as well as in the type of the bond between glucosamine and rhamnose residues (1-->2 or 1-->3).
Sigma-Aldrich
2-Hydroxyhexadecanoic acid, ≥98% (capillary GC)