Merck
  • Addition of malonyl radicals to glycals with C-1 acceptor groups: remarkable influence of the substituents on the product distribution.
Addition of malonyl radicals to glycals with C-1 acceptor groups: remarkable influence of the substituents on the product distribution.
Chemical communications (Cambridge, England) (2002-07-12)
Viktor Gyóllai, Dirk Schanzenbach, László Somsák, Torsten Linker
ABSTRACT
The ceric(IV) ammonium nitrate (CAN)-mediated radical addition of dimethyl malonate to glycals 1 affords methyl glycosides 2 and ortho esters 3 as main products; the product distribution strongly depends on the substitution pattern at the 1-position, which can be rationalized in terms of the oxidation potentials of the intermediary anomeric radicals.