Merck
  • A novel isomerization on interaction of antitumor-active azole-bridged dinuclear platinum(II) complexes with 9-ethylguanine. Platinum(II) atom migration from N2 to N3 on 1,2,3-triazole.
A novel isomerization on interaction of antitumor-active azole-bridged dinuclear platinum(II) complexes with 9-ethylguanine. Platinum(II) atom migration from N2 to N3 on 1,2,3-triazole.
Journal of the American Chemical Society (2002-04-25)
Seiji Komeda, Martin Lutz, Anthony L Spek, Yasuyuki Yamanaka, Takaji Sato, Masahiko Chikuma, Jan Reedijk
ABSTRACT
The reactions of the dinuclear platinum(II) complexes, [[cis-Pt(NH(3))(2)](2)(mu-OH)(mu-pz)](NO(3))(2) (1, pz = pyrazolate), [[cis-Pt(NH(3))(2)](2)(mu-OH)(mu-1,2,3-ta-N1,N2)](NO(3))(2) (2, 1,2,3-ta = 1,2,3-triazolate), and a newly prepared [[cis-Pt(NH(3))(2)](2)(mu-OH)(mu-4-phe-1,2,3-ta-N1,N2)](NO(3))(2) (3, 4-phe-1,2,3-ta = 4-phenyl-1,2,3-triazolate), whose crystal structure was determined, with 9-ethylguanine (9EtG) have been monitored in aqueous solution at 310 K by means of (1)H NMR spectroscopy. The dinuclear platinum(II) complexes 1-3 each react with 9EtG in a bifunctional way to form 1:2 complexes, [[cis-Pt(NH(3))(2)(9EtG-N7)](2)(mu-pz)](3+) (4), [[cis-Pt(NH(3))(2)(9EtG-N7)](2)(mu-1,2,3-ta-N1,N3)](3+) (5), and [[cis-Pt(NH(3))(2)(9EtG-N7)](2)(mu-4-phe-1,2,3-ta-N1,N3)](3+) (6). The reactions of 2 and 3 involve a novel isomerization, in which the Pt atom, initially bound to N2 on the 1,2,3-ta, migrates to N3 after the first substitution by N7 of 9EtG. This isomerization reaction has been unambiguously characterized by 1D and 2D NMR spectroscopy and pH titration. The reactions of 2 and 3 with 9EtG show faster kinetics, and the second-order rate constants (k) for the reactions of 1-3are 1.57 x 10(-4), 2.53 x 10(-4), and 2.56 x 10(-4) M(-1) s(-1), respectively. The pK(a) values at the N1H site of 9EtG were determined for 4-6 from the pH titration curves. Cytotoxicity assays of 1-3 were performed in L1210 murine leukemia cell lines, respectively sensitive and resistant to cisplatin. In the parent cell line, 2 and 3 exhibit higher cytotoxicity compared to cisplatin, especially, 2 is 10 times as active as cisplatin. 1 was found to be less cytotoxic than cisplatin, but still in the active range and more active than cisplatin in a cisplatin-resistant cell line.
Sigma-Aldrich
9-Ethylguanine