Merck
  • Recombinant human tumor necrosis factor alpha does not potentiate cell killing after photodynamic therapy with a silicon phthalocyanine in A431 human epidermoid carcinoma cells.
Recombinant human tumor necrosis factor alpha does not potentiate cell killing after photodynamic therapy with a silicon phthalocyanine in A431 human epidermoid carcinoma cells.
International journal of oncology (2001-02-15)
K Azizuddin, K Kalka, S M Chiu, N Ahmad, H Mukhtar, D Separovic
ABSTRACT
Photodynamic therapy (PDT) is a novel cancer treatment utilizing a photosensitizer, visible light and oxygen. PDT with the silicon phthalocyanine Pc 4, a new photosensitizer, is highly effective in cancer cell destruction and tumor ablation. The mechanisms underlying cancer cell killing by PDT are not fully understood. Tumor necrosis factor alpha (TNF) is a multifunctional cytokine that has been implicated in photocytotoxicity. We asked whether recombinant human TNF (rhTNF) affects Pc 4-PDT cytotoxicity in A431 human epidermoid carcinoma cells. Co-treatment of A431 cells with various doses of Pc 4-PDT and a sub-lethal rhTNF dose led to a sub-additive reduction in cell survival. In addition, in the presence of Pc 4-PDT or rhTNF, caspase-3 activity and apoptosis were induced. The combined treatment, however, did not potentiate either caspase-3 activity or apoptosis. Similar to previous findings we observed that Pc 4-PDT initiated a time-dependent extracellular TNF accumulation. The data suggest that: a) PDT and rhTNF induce cancer cell killing through different mechanisms; and b) Pc 4-PDT-induced TNF production is a stress response that may not directly affect photocytotoxicity.