Merck
  • Efficacy and safety of flupirtine maleate and tramadol hydrochloride in postoperative pain management--a prospective randomised double blinded study.
Efficacy and safety of flupirtine maleate and tramadol hydrochloride in postoperative pain management--a prospective randomised double blinded study.
Journal of the Indian Medical Association (2012-10-04)
Syed Mohammed Naser, Niladri Sarkar, Arunava Biswas, Firdaus Kamal, Raghu Prakash, Q M Rahaman, Anup Kumar Das
ABSTRACT
The study was conducted to evaluate the efficacy and safety of flupirtine maleate 100 mg thrice daily compared to tramadol hydrochloride 50 mg thrice daily as postoperative pain management for 5 days. A total of 113 postoperative patients were recruited for the study. Those who met the inclusion criteria (n = 104) were randomised into two treatment groups. One of the groups received flupirtine maleate and the other tramadol hydrochloride both orally. The pain intensity was assessed by visual analogue scale. Patients were informed to report any adverse effect encountered during the study period. The overall effect of the drug (global assessment of the study medication) on pain and side-effects was assessed by the patients at the end of the trial on a categorical scale. There was significant reduction in pain score (p < 0.001) in the flupirtine group with almost equal efficacy to that of tramadol group but the incidence of adverse effects were much less (7.4%) and didn't need discontinuation of the study. All drugs were assessed as good. Therefore it can be concluded that oral flupirtine can deliver the same analgesic efficacy as oral tramadol for postoperative pain relief, which might be beneficial for avoiding the adverse effects ofopioids and non-steroidal anti-inflammatory drug therapy.
Sigma-Aldrich
Flupirtine maleate salt, ≥98% (HPLC)