Merck
  • Allergen-induced recruitment of Fc epsilon RI+ eosinophils in human atopic skin.
Allergen-induced recruitment of Fc epsilon RI+ eosinophils in human atopic skin.
European journal of immunology (1997-05-01)
L T Barata, S Ying, J A Grant, M Humbert, J Barkans, Q Meng, S R Durham, A B Kay
ABSTRACT
We have attempted to identify Fc epsilon RI+ eosinophils in cutaneous late-phase reaction in atopic subjects biopsied at 6, 24 and 48 h after the injection of either allergen or a diluent control. Compared to the diluent sites, allergen-injected sites had significantly increased numbers of eosinophils, peaking between 6 and 24 h, of which approximately 20-30% expressed mRNA for the alpha, beta, and gamma chains of Fc epsilon RI, as shown by in situ hybridization. Using either a monoclonal or a polyclonal anti-alpha chain antibody, the Fc epsilon RI alpha protein also co-localized to approximately 50-80% of eosinophils at all time points studied. We also observed a significant correlation (r = 0.89; p = 0.02) between the numbers of Fc epsilon RI+ (997+)/EG2+ eosinophils and the magnitude of the late-phase reaction. Thus, a significant proportion of eosinophils infiltrating the site of allergen-induced allergic tissue reactions in atopic subjects express Fc epsilon. RI. The findings show that high-affinity IgE receptors may play a role in eosinophil secretory processes in vivo.