Merck
  • The cardiovascular and renal effects of hydrins and arginine vasotocin in frogs.
The cardiovascular and renal effects of hydrins and arginine vasotocin in frogs.
General and comparative endocrinology (1993-07-01)
K W Chiu, Y C Lee, P K Pang
ABSTRACT
Cardiovascular and renal actions of hydrins and arginine vasotocin (AVT) in frogs (Rana tigrina and Rana catesbeiana) were examined in view of their relationships as precursor/prohormone-hormones in the anuran amphibian. In vitro hydrins and AVT respectively vasorelaxed and vasoconstricted KCl-preconstricted femoral artery in R. tigrina. AVT also produced dose-dependent contractions on untreated preparations; hydrins had no effect on these preparations. Both the hydrins and AVT were positively inotropic and chronotropic, with AVT being more potent. In vivo hydrins and AVT were anti-diuretic except at high doses, when AVT became diuretic. The hydrins and AVT were vasodepressor and pressor, respectively, in R. catesbeiana. These data suggest that the addition of one to three amino acids onto the AVT molecule reverses the contractile response in the vascular smooth muscle. With this dissociation of the vascular effect from that of the renal, the hydrins might play a physiological role in water balance of frogs.