- Detection and identification of famprofazone and its metabolite in human urine.
Detection and identification of famprofazone and its metabolite in human urine.
Journal of chromatography. B, Biomedical applications (1994-11-18)
H S Shin, J S Park, P B Park, S J Yun
PMID7894665
ABSTRACT
Detection and identification of famprofazone and its metabolite, p-hydroxydesmethylfamprofazone, were described. The identity of the new metabolite was confirmed by comparing its mass spectrum and gas chromatographic retention time with that of the synthetic standard and its derivatives. Unchanged famprofazone was detected up to 6 h and the metabolite was detected up to 32 h in human urine after administration of famprofazone. The sum of the two compounds excreted in urine was ca. 1.5% of the dose.