Merck
  • A comparative assessment of efficacy of three anticoagulant rodenticides.
A comparative assessment of efficacy of three anticoagulant rodenticides.
Journal of hygiene, epidemiology, microbiology, and immunology (1982-01-01)
D M Renapurkar
ABSTRACT
Results are presented of feeding tests carried out with three common anticoagulant rodenticides viz., coumatetralyl, fumarin and warfarin on three common species of commensal rodents i.e., Rattus rattus, Rattus norvegicus and Bandicota bengalensis. All three species of rodents were susceptible to anticoagulant rodenticides. However, the action of these compounds in B. bengalensis was comparatively slow. Coumatetralyl was found to be the most effective rodenticide followed by fumarin and warfarin. Liquid baits of these compounds are more effective in comparison to food baits.
Product Description
Supelco
Coumatetralyl, PESTANAL®, analytical standard