Merck
  • Synthesis of substituted indoline and carbazole by benzyne-mediated cyclization-functionalization.
Synthesis of substituted indoline and carbazole by benzyne-mediated cyclization-functionalization.
Organic letters (2013-04-04)
Toshiharu Noji, Hideto Fujiwara, Kentaro Okano, Hidetoshi Tokuyama
ABSTRACT
A benzyne-mediated synthesis of substituted indolines and carbazoles was developed. The reaction includes generation of benzyne using Mg(TMP)2·2LiCl as a base, cyclization, and trapping the resulting organomagnesium intermediate with an electrophile to provide a series of substituted indolines and carbazoles in a regiospecific manner. This was applied to a concise five-pot total synthesis of heptaphylline.
Sigma-Aldrich
Indoline, ReagentPlus®, 99%