Merck
  • The role of 14-3-3ε interaction with phosphorylated Cdc25B at its Ser321 in the release of the mouse oocyte from prophase I arrest.
The role of 14-3-3ε interaction with phosphorylated Cdc25B at its Ser321 in the release of the mouse oocyte from prophase I arrest.
PloS one (2013-01-18)
Jun Meng, Cheng Cui, Yanchun Liu, Minglin Jin, Didi Wu, Chao Liu, Enhua Wang, Bingzhi Yu
ABSTRACT
The protein kinase A (PKA)/Cdc25B pathway plays a critical role in maintaining meiotic arrest in mouse oocytes. However, the molecular mechanism underlying this interchange is not known. In this study, we assessed the role of 14-3-3ε interaction with phosphorylated Cdc25B at its Ser321 as the mouse oocyte is released from prophase I arrest. The 14-3-3ε isoform is a highly conserved protein with various regulatory roles, including maintenance of meiotic arrest. Cdc25B phosphatase is also a key cell cycle regulator. 14-3-3ε binds to Cdc25B-WT, which was abrogated when Ser321 of Cdc25B was mutated to Ala. In addition, we found that 14-3-3ε and Cdc25B were co-localized. Cdc25B was translocated from the cytoplasm to the nucleus shortly before germinal vesicle breakdown (GVBD) during the primary oocyte stage of oogenesis. However, mutation of Ser321 to Ala completely abolished the cytoplasmic localization of Cdc25B. Furthermore, oocytes co-expressing of Cdc25B-WT or Cdc25B-Ser321D and 14-3-3ε were unable to undergo GVBD. In contrast, co-expression of 14-3-3ε and Cdc25B-Ser321A induced GVBD and allowed the process to continue. Down-regulation of 14-3-3ε caused partial meiotic resumption. Taken together, these data indicate that Ser321 of Cdc25B is the specific binding site for 14-3-3ε binding, and that 14-3-3ε is the significant factor in Cdc25B regulation during meiotic resumption of GV stage.
Sigma-Aldrich
O-Phospho-DL-serine, ≥98.0% (NT)