Merck
  • UV-cross-linkable multilayer microcapsules made of weak polyelectrolytes.
UV-cross-linkable multilayer microcapsules made of weak polyelectrolytes.
Langmuir : the ACS journal of surfaces and colloids (2012-06-27)
Qiangying Yi, Dongsheng Wen, Gleb B Sukhorukov
ABSTRACT
Microcapsules composed of weak polyelectrolytes modified with UV-responsive benzophenone (BP) groups were fabricated by the layer-by-layer (LbL) technique. Being exposed to UV lights, capsules shrunk in the time course of minutes at irradiation intensity of 5 mW/cm(2). The shrinkage adjusted the capsule permeability, providing a novel way to encapsulate fluorescence-labeled dextran molecules without heating. Cross-linking within the capsule shells based on hydrogen abstraction via excited benzophenone units by UV showed a reliable and swift approach to tighten and stabilize the capsule shell without losing the pH-responsive properties of the weak polyelectrolyte multilayers.