Merck
  • Retrograde signaling by the plastidial metabolite MEcPP regulates expression of nuclear stress-response genes.
Retrograde signaling by the plastidial metabolite MEcPP regulates expression of nuclear stress-response genes.
Cell (2012-06-26)
Yanmei Xiao, Tatyana Savchenko, Edward E K Baidoo, Wassim E Chehab, Daniel M Hayden, Vladimir Tolstikov, Jason A Corwin, Daniel J Kliebenstein, Jay D Keasling, Katayoon Dehesh
ABSTRACT
Plastid-derived signals are known to coordinate expression of nuclear genes encoding plastid-localized proteins in a process termed retrograde signaling. To date, the identity of retrograde-signaling molecules has remained elusive. Here, we show that methylerythritol cyclodiphosphate (MEcPP), a precursor of isoprenoids produced by the plastidial methylerythritol phosphate (MEP) pathway, elicits the expression of selected stress-responsive nuclear-encoded plastidial proteins. Genetic and pharmacological manipulations of the individual MEP pathway metabolite levels demonstrate the high specificity of MEcPP as an inducer of these targeted stress-responsive genes. We further demonstrate that abiotic stresses elevate MEcPP levels, eliciting the expression of the aforementioned genes. We propose that the MEP pathway, in addition to producing isoprenoids, functions as a stress sensor and a coordinator of expression of targeted stress-responsive nuclear genes via modulation of the levels of MEcPP, a specific and critical retrograde-signaling metabolite.
Sigma-Aldrich
meso-Erythritol, ≥99% (GC)
Pricing and availability is not currently available.