Merck
  • Cellulose acetate electrospun fiber mats for controlled release of silymarin.
Cellulose acetate electrospun fiber mats for controlled release of silymarin.
Journal of nanoscience and nanotechnology (2012-04-25)
Manisara Phiriyawirut, Thawatchai Phaechamud
ABSTRACT
In this research, the silymarin-loaded electrospun cellulose acetate (CA) fibers were prepared which containing silymarin in various amounts (i.e., 2.5-20 wt.% based on the weight of CA powder). Incorporation of silymarin in the neat CA solution did not affect the morphology of the resulting fibers, as both the neat and the silymarin-loaded CA fibers were smooth. The average diameters of silymarin-loaded CA fiber ranged between 550-900 nm. No presence of the silymarin aggregates of any kind was observed on the surfaces of these fibers, suggesting that the silymarin was encapsulated well within the fibers. These results were confirmed by lowering the glass transition temperature and the melting temperature of the silymarin-loaded electrospun CA fibers which is determined by DSC technique. The release characteristic of silymarin from the silymarin-loaded CA fiber mats was investigated by the total immersion in the solution of 1/1 phosphate buffer/methanol medium pH 7.4 at 37 degrees C. The silymarin release from the silymarin-loaded electrospun CA fiber mat is monotonously increased to reach the maximum value at 480 min. The maximum amount of silymarin released from these materials increases with the increasing of initial silymarin loading in the spinning CA solutions. Since no aggregation of silymarin was found on the surface of the silymarin-loaded fibers, the release of the silymarin from fiber mats was mainly by the diffusion.