Merck
  • Design of xanthone propionate photolabile protecting group releasing acyclovir for the treatment of ocular herpes simplex virus.
Design of xanthone propionate photolabile protecting group releasing acyclovir for the treatment of ocular herpes simplex virus.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2012-01-10)
Jessie A Blake, Bettina Bareiss, Liliana Jimenez, May Griffith, J C Scaiano
ABSTRACT
We have attached the antiviral drug acyclovir (ACV) to a xanthone photolabile protecting group (or photocage) through the O6 position of acyclovir, a procedure designed for the treatment of ocular herpes simplex virus infections. Acyclovir is photoreleased from the photocage, under physiological conditions, with a quantum yield (Φ(ACV release)) of 0.1-0.3 and an uncaging cross section (Φ·ε) of 450-1350 M cm(-1). We demonstrate that this photorelease method outcompetes alternative reaction pathways, such as protonation. Furthermore, complete release of the drug is theoretically possible given a sufficient dose of light . Surprisingly the acyclovir photocage, also showed some antiviral activity towards HSV-1.
Sigma-Aldrich
Xanthone, 97%