Merck
  • Hereditary fructose intolerance: frequency and spectrum mutations of the aldolase B gene in a large patients cohort from France--identification of eight new mutations.
Hereditary fructose intolerance: frequency and spectrum mutations of the aldolase B gene in a large patients cohort from France--identification of eight new mutations.
Molecular genetics and metabolism (2008-06-11)
Anne Davit-Spraul, Catherine Costa, Mokhtar Zater, Dalila Habes, Jacques Berthelot, Pierre Broué, François Feillet, Olivier Bernard, Philippe Labrune, Christiane Baussan
ABSTRACT
We investigated the molecular basis of hereditary fructose intolerance (HFI) in 160 patients from 92 families by means of a PCR-based mutation screening strategy, consisting of restriction enzyme digestion and direct sequencing. Sixteen different mutations of the aldolase B (ALDOB) gene were identified in HFI patients. As in previous studies, p.A150P (64%), p.A175D (16%) and p.N335K (5%) were the most common mutated alleles, followed by p.R60X, p.A338V, c.360_363delCAAA (p.N120KfsX30), c.324G>A (p.K108K) and c.625-1G>A. Eight novel mutations were also identified in 10 families with HFI: a one-base deletion (c.146delT (p.V49GfsX27)), a small deletion (c.953del42bp), a small insertion (c.689ins TGCTAA (p.K230MfsX136)), one splice site mutation (c.112+1G>A), one nonsense mutation (c.444G>A (p.W148X)), and three missense mutations (c.170G>C (p.R57P), c.839C>A (p.A280P) and c.932T>C (p.L311P)). Our strategy allows to diagnose 75% of HFI patients using restriction enzymatic analysis and to enlarge the diagnosis to 97% of HFI patients when associated with direct sequencing.