Merck
  • Influence of bestatin, an inhibitor of aminopeptidases, on T and B lymphocyte subsets in mice.
Influence of bestatin, an inhibitor of aminopeptidases, on T and B lymphocyte subsets in mice.
Polish journal of veterinary sciences (2011-10-01)
M Lis, M Szczypka, A Suszko, B Obmińska-Mrukowicz
ABSTRACT
Bestatin, a low-molecular weight dipeptide, is a potent inhibitor of aminopeptidase N which has been demonstrated to have antitumor and immunomodulatory effects. The effects of bestatin (10, 1 and 0.1 mg/kg) administered intraperitoneally once, five or ten times to mice on the total number of lymphocytes in the thymus, spleen and mesenteric lymph nodes and the percentage and the absolute number of T cell subsets (CD4+CD8+, CD4-CD8-, CD4+, CD8+) in the thymus and T (CD3+, CD4+, CD8+) and B (CD19+) lymphocytes in the spleen and mesenteric lymph nodes were studied. It has been found that bestatin administered ten times at doses of 10, 1 and 0.1 mg/kg increased the total number of thymocytes, splenocytes and lymphocytes of mesenteric lymph nodes. Bestatin also changed the percentage and the absolute number of T cell subsets in the thymus and T and B lymphocytes in the peripheral lymphatic organs. Five and ten exposures to bestatin (10, 1 and 0.1 mg/kg) increased the absolute count of both immature CD4+CD8+ and CD4-CD8- thymic cells. Moreover, both a single and multiple administration of bestatin (1 and 0.1 mg/kg) decreased the percentage and absolute count of CD3+ splenocytes and mesenteric lymph node cells with corresponding decreases in the percentage and absolute count of CD4+ and CD8+ cells. Both a single and multiple administration of bestatin at all the doses under investigation augmented the percentage and the absolute count of CD19+ (B lymphocytes) in the peripheral lymphatic organs. The results of the study show that there is a relationship between the effect induced by bestatin and the dose of the drug as well as the number of doses applied. The strongest effect on the T and B lymphocyte subsets was noted after five injections of bestatin at doses of 1 and 0.1 mg/kg.
Sigma-Aldrich
N-[(2S,3R)-3-Amino-2-hydroxy-4-phenylbutyryl]-L-leucine, 97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Bestatin hydrochloride, ≥98% (HPLC)