Merck
  • Targeting glycolysis: a fragment based approach towards bifunctional inhibitors of hLDH-5.
Targeting glycolysis: a fragment based approach towards bifunctional inhibitors of hLDH-5.
Chemical communications (Cambridge, England) (2010-08-03)
Adam D Moorhouse, Christian Spiteri, Pallavi Sharma, Mire Zloh, John E Moses
ABSTRACT
hLDH-5 has emerged as a promising target for anti-glycolytic cancer chemotherapy. Here we report a first generation of bifunctional inhibitors, which show promising activity against hLDH-5.
Sigma-Aldrich
Oxamic acid, ≥98%