- Wheat (Triticum aestivum L.) response to a zinc fertilizer applied as zinc lignosulfonate adhered to a NPK fertilizer.
Wheat (Triticum aestivum L.) response to a zinc fertilizer applied as zinc lignosulfonate adhered to a NPK fertilizer.
The efficacy as Zn fertilizers for wheat of zinc lignosulfonate (ZnLS) products adhered to NPK was evaluated by three plant experimental designs. In the first and second assays, wheat plants were grown under controlled conditions with perlite and a calcareous soil as substrate, respectively. Shoot dry matter and Zn concentration showed that NPK + ZnLS was a better Zn source for wheat than NPK + ZnSO(4) under our experimental conditions. A third experiment was conducted under field conditions on a calcareous soil with a low Zn level. Wheat samples were taken at five growth stages of the crop. Although at early stages NPK + ZnLS was the most efficient source of Zn, at harvest no significant differences among treatments were found. Despite that, NPK + ZnLS showed evidence of being a useful Zn source for wheat crop under calcareous conditions.