Merck
Telephone-based management of lactation mastitis.
The Journal of reproductive medicine (2009-06-13)
Wendy L Berth, Charles W Schauberger, Mary A Alvarado, Michelle A Mathiason
ABSTRACT
To evaluate the quality of care for mastitis managed by telephone without an office or emergency room visit. A word search of electronic medical records allowed for identification of patients managed with a mastitis clinical practice guideline. Comparisons of outcomes were made to a group of patients who received care in the clinic, urgent care or emergency room. Rates of successful resolution with 1 encounter were 86% for telephone-based care and 82% for visit-based care (p = 0.550). Antibiotic selection was enhanced by use of the standing order. Telephone-based management of mastitis may be safe, effective, and efficient.
Sigma-Aldrich
Dicloxacillin sodium salt monohydrate
Pricing and availability is not currently available.