Merck
  • Purification and characterization of human brain serine racemase expressed in moderately halophilic bacteria.
Purification and characterization of human brain serine racemase expressed in moderately halophilic bacteria.
Protein and peptide letters (2009-02-10)
Chizuru Nagayoshi, Matsujiro Ishibashi, Masao Tokunaga
ABSTRACT
We have successfully expressed an active human brain serine racemase (hSR) with His-tag using moderate halophile. The purified His-hSR showed high elimination and racemization activities on L-serine: the elimination activity was 2.6-fold higher than racemization activity. Both enzyme activities showed an optimum reaction pH at around 9.0 and were stimulated 5- to 7-fold by such divalent cations as Mg++, Mn++ and Ca++.