Merck
  • Povarov reactions involving 3-aminocoumarins: synthesis of 1,2,3,4-tetrahydropyrido[2,3-c]coumarins and pyrido[2,3-c]coumarins.
Povarov reactions involving 3-aminocoumarins: synthesis of 1,2,3,4-tetrahydropyrido[2,3-c]coumarins and pyrido[2,3-c]coumarins.
The Journal of organic chemistry (2008-09-30)
Amit A Kudale, Jamie Kendall, David O Miller, Julie L Collins, Graham J Bodwell
ABSTRACT
Condensation of 3-aminocoumarin (5) with 4-nitrobenzaldehyde (8) afforded a 2-azadiene (9), which reacted with various electron-rich alkenes (10 examples) in the presence of Yb(OTf)3 to afford 1,2,3,4-tetrahydropyrido[2,3-c]coumarins. Yields were generally good, but the diastereomeric ratios were highly variable. The products arose through a formal [4 + 2] cycloaddition (inverse electron demand Diels-Alder reaction) followed by tautomerization. As such, these are examples of the Povarov reaction. A range of 1,2,3,4-tetrahydropyrido[2,3-c]coumarins was then synthesized using a three-component version of this reaction, which involves in situ formation of the 2-azadiene component. Some of these products were converted into the corresponding pyrido[2,3-c]coumarins upon treatment with various oxidants, the most effective of which proved to be nitrous gases.
Sigma-Aldrich
3-Aminocoumarin, 97%
Sigma-Aldrich
4-Nitrobenzaldehyde, 98% (GC)