Merck
  • Enantioselective intramolecular hydroarylation of alkenes via directed C-H bond activation.
Enantioselective intramolecular hydroarylation of alkenes via directed C-H bond activation.
The Journal of organic chemistry (2008-08-07)
Hitoshi Harada, Reema K Thalji, Robert G Bergman, Jonathan A Ellman
ABSTRACT
Highly enantioselective catalytic intramolecular ortho-alkylation of aromatic imines containing alkenyl groups tethered at the meta position relative to the imine directing group has been achieved using [RhCl(coe)2]2 and chiral phosphoramidite ligands. Cyclization of substrates containing 1,1- and 1,2-disubstituted as well as trisubstituted alkenes were achieved with enantioselectivities >90% ee for each substrate class. Cyclization of substrates with Z-alkene isomers proceeded much more efficiently than substrates with E-alkene isomers. This further enabled the highly stereoselective intramolecular alkylation of certain substrates containing Z/E-alkene mixtures via a Rh-catalyzed alkene isomerization with preferential cyclization of the Z-isomer.