Merck
A novel technique to perform direct intraganglionar injections in rats.
Journal of neuroscience methods (2006-09-16)
L F Ferrari, F Q Cunha, C A Parada, S H Ferreira
ABSTRACT
The present work describes a simple method for direct drug administration into the dorsal root ganglion (DRG) in anesthetized rats. This technique does not involve surgery, is easy to learn and allows behavioral testing within minutes after the injection. Based on landmarks that target the L5 DRG, an orifice was created with a guide needle through which a specially designed needle was inserted for solution injection. Its introduction into the ganglia was ensured by the triggering of an ipsilateral hindpaw reflex. The precision of the technique was checked by injections of the biological dye Pontamine Sky Blue (PSB) or C14-labeled arginine. There was no leakage of the dye to the surrounding tissues after a single 4 microl or three successive 2.5 microl injections (at 30-min intervals). Moreover, identical effects were observed with prostaglandin E2 (PGE2), morphine or glibenclamide injected intraplantarly or in the DRG, thus confirming the precision of the method and suggesting that the ganglion cells and peripheral nociceptors may display similar receptor population.