Merck
  • Efficient terpene hydroxylation catalysts based upon P450 enzymes derived from actinomycetes.
Efficient terpene hydroxylation catalysts based upon P450 enzymes derived from actinomycetes.
Organic & biomolecular chemistry (2005-09-28)
Ayhan Celik, Sabine L Flitsch, Nicholas J Turner
ABSTRACT
The hydroxylation of alpha-ionone 1 and beta-ionone 2 to their corresponding mono-hydroxylated derivatives has been examined using a recombinant E. coli whole cell system, in which cytochromes P450 SU1 and SU2, and P450 SOY were over-expressed with their cognate ferrodoxins. Both substrates are hydroxylated with a high degree of regioselectivity and for alpha-ionone 1 the reaction is highly diastereoselective yielding the anti-isomer.
Sigma-Aldrich
α-Ionone, ≥90%, stabilized
Sigma-Aldrich
α-Ionone, technical grade, 90%