Merck
  • Natural occurrence of trichothecenes on lodged and water-damaged domestic rice in Japan.
Natural occurrence of trichothecenes on lodged and water-damaged domestic rice in Japan.
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan (2004-07-27)
Kenji Tanaka, Hidetaka Kobayashi, Tadahiro Nagata, Masaru Manabe
ABSTRACT
In 1998, a typhoon struck before rice harvesting in Japan, and the unpolished rice was found to be stained brown. Samples were collected and analyzed for the presence of trichothecenes using GC/MS. The mycotoxins deoxynivalenol (DON), fusarenon-X (Fus.-X) and nivalenol (NIV) were detected and confirmed with GC/MS. The quantity of trichothecenes was determined using GC-ECD. To our knowledge, this is the first report of the presence of the trichothecene Fus.-X in rice.
Product Description
Supelco
Fusarenon X solution, 100 μg/mL in acetonitrile, analytical standard
Supelco
Fusarenon X, analytical standard