Merck
  • Exceptional chiral recognition of racemic carboxylic acids by calix[4]arenes bearing optically pure alpha,beta-amino alcohol groups.
Exceptional chiral recognition of racemic carboxylic acids by calix[4]arenes bearing optically pure alpha,beta-amino alcohol groups.
Organic letters (2004-04-09)
Yan-Song Zheng, Chun Zhang
ABSTRACT
Calixarenes bearing optically pure alpha,beta-amino alcohol groups at their lower rim exhibit exceptional and efficient chiral recognition ability in discrimination of racemic mandelic acid, 2,3-dibenzoyltartaric acid and 2-hydroxy-3-methylbutyric acid. [structure: see text]
Sigma-Aldrich
Dibenzoyl-L-tartaric acid, 98%
Sigma-Aldrich
(+)-2,3-Dibenzoyl-D-tartaric acid, ≥99.0% (T)
Sigma-Aldrich
(+)-2,3-Dibenzoyl-D-tartaric acid, ≥98%, made from synthetic tartaric acid