Merck
  • Simultaneous biodegradation of a phenol and 3,4-dimethylphenol mixture under denitrifying conditions.
Simultaneous biodegradation of a phenol and 3,4-dimethylphenol mixture under denitrifying conditions.
Water science and technology : a journal of the International Association on Water Pollution Research (2003-12-03)
L Puig-Grajales, O Rodríguez-Nava, E Razo-Flores
ABSTRACT
Denitrification is a feasible alternative for the treatment of phenolic bearing-wastewaters. The aim of this study was to evaluate the biodegradability of phenolic compounds, as the only carbon and energy source in batch and continuous experiments, using nitrate as a final electron acceptor. Experiments in a continuous upward anaerobic sludge bed reactor demonstrated the possibility of biodegrading a mixture of phenol and 3,4-dimethylphenol at organic loads of 251.6 and 39.5 mg/L-d, respectively, at a COD/NO3(-)-N ratio of 2.57. A nitrogen production efficiency of 86% was obtained according to the nitrate consumption. GC-MS analyses demonstrated that m-cresol was an intermediate of 3,4-dimethylphenol degradation in batch conditions, and had an inhibitory effect on phenol degradation.
Sigma-Aldrich
3,4-Dimethylphenol, 98%