- Coimmobilization of urease and glutamate dehydrogenase in electrochemically prepared polypyrrole-polyvinyl sulfonate films.
Coimmobilization of urease and glutamate dehydrogenase in electrochemically prepared polypyrrole-polyvinyl sulfonate films.
Applied biochemistry and biotechnology (2002-01-11)
A Gambhir, M Gerard, A K Mulchandani, B D Malhotra
PMID11783891
ABSTRACT
Immobilization of urease and glutamate dehydrogenase enzymes in electrochemically prepared polypyrrole-polyvinyl sulfonate films (PPY-PVS) was carried out using physical adsorption and electrochemical entrapment techniques. Detailed studies on optimum pH, Fourier transform infrared spectroscopy, cyclic voltammetry, and scanning electron microscopy of the enzymes in the immobilized state were conducted. The value of the apparent Michaelis-Menten constant was experimentally determined to be 2.5 and 2.7 for physically adsorbed and electrochemically entrapped urease in PPY-PVS films, respectively.