- Rapid degradation of ferulic acid via 4-vinylguaiacol and vanillin by a newly isolated strain of bacillus coagulans.
Rapid degradation of ferulic acid via 4-vinylguaiacol and vanillin by a newly isolated strain of bacillus coagulans.
Journal of biotechnology (2000-08-19)
B Karmakar, R M Vohra, H Nandanwar, P Sharma, K G Gupta, R C Sobti
PMID10949310
ABSTRACT
A new strain Bacillus coagulans BK07 was isolated from decomposed wood-bark, based on its ability to grow on ferulic acid as a sole carbon source. This strain rapidly decarboxylated ferulic acid to 4-vinylguaiacol, which was immediately converted to vanillin and then oxidized to vanillic acid. Vanillic acid was further demethylated to protocatechuic acid. Above 95% substrate degradation was obtained within 7 h of growth on ferulic acid medium, which is the shortest period of time reported to date. The major degradation products, was isolated and identified by thin-layer chromatography, high performance liquid chromatography and 1H-nuclear magnetic resonance spectroscopy were 4-vinylguaiacol, vanillin, vanillic acid and protocatechuic acid.